Platinum : rhodium (90:10) (w/w), wire Ø 0.25 mm for ISA type R standard grade thermocouple
Supplier: BeanTown Chemical
219070-100CM
219070-25CM
219070-500CM
BT219070-100CMEA
264
USD
BT219070-100CM
BT219070-25CM
BT219070-500CM
Platinum : rhodium (90:10) (w/w), wire Ø 0.25 mm for ISA type R standard grade thermocouple
Platinum : rhodium
MDL No: MFCD00198186
Solid; Formula: Pt:Rh; 90:10 wt%
- Solid