Hafnium(IV) tert-butoxide ≥99.9%
Supplier: BeanTown Chemical
Synonyms:
Hafnium(IV) t-butoxide, Hafnium(IV)-tert-butanolate
219065-5G
BT219065-1GEA
80.3
USD
BT219065-1G
BT219065-5G
Hafnium(IV) tert-butoxide ≥99.9%
Hafnium(IV) tert-butoxide
CAS: 2172-02-3; MDL No: MFCD00070458
Liquid; Linear Formula: Hf[OC(CH3)3]4 ; Molecular Formula: C16H36HfO4; MW: 470.65
Boiling Point: 90°/5 mmHg; Flash point: 28°C (82°F)
Density (g/mL): 1.166; Refractive Index: 1.424
Moisture Sensitive
- Refractive index: 1.424
- Moisture Sensitive
- Liquid
Formula:
C₁₆H₃₆HfO₄ Boiling Pt: 90 °C/5 mmHg Density: 1.166 g/mL Flash Pt: 28 °C (82 °F) |
MDL Number:
MFCD00070458 CAS Number: 2172-02-3 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...