Tellurium dioxide ≥99.99%
Supplier: BeanTown Chemical
218820-100G
218820-25G
BT218820-100GEA
261.03
USD
BT218820-100G
BT218820-25G
Tellurium dioxide ≥99.99%
Tellurium dioxide
CAS: 7446-07-3; EC No: 231-193-1; MDL No: MFCD00011263; RTECS: WY2675000
Powder; Linear Formula: TeO2; MW: 159.60
Melting Point: 733°
Density (g/mL): 5.67
- RTECS: WY2675000
- Powder
Formula:
O₂Te Melting Pt: 733 °C Density: 5.67 g/mL |
MDL Number:
MFCD00011263 CAS Number: 7446-07-3 EINECS: 231-193-1 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...