4-Fluoro-3-phenoxybenzaldehyde ≥98%
Supplier: BeanTown Chemical
218650-25G
218650-5G
BT218650-25GEA
209
USD
BT218650-25G
BT218650-5G
4-Fluoro-3-phenoxybenzaldehyde ≥98%
4-Fluoro-3-phenoxybenzaldehyde
CAS: 68359-57-9; EC No: 269-856-2; MDL No: MFCD01318148
UN No: UN3082; Haz Class: 9; Packing Group: III
Liquid; Molecular Formula: C13H9FO2; MW: 216.21
Density (g/mL): 1.2091; Refractive Index: 1.583
- Refractive index: 1.583
- Liquid
Formula:
C₁₃H₉FO₂ MW: 216.21 g/mol Density: 1.2091 g/mL |
MDL Number:
MFCD01318148 CAS Number: 68359-57-9 EINECS: 269-856-2 ADR: 9 III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...