4-Bromopyridinium chloride ≥99%

Supplier: BeanTown Chemical

Synonyms: 4-Bromopyridine HCl, 4-Bromopyridine hydrochloride

218605-1G 218605-25G 218605-5G
BT218605-1GEA 34 USD
BT218605-1G BT218605-25G BT218605-5G
4-Bromopyridinium chloride ≥99%
4-Bromopyridinium chloride

CAS: 19524-06-2; EC No: 243-128-4; MDL No: MFCD00012828 Crystalline; Molecular Formula: C5H4BrN·HCl ; MW: 194.46 Melting Point: 270° (decomposes) Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: C₅H₅BrClN
Melting Pt: 270 °C (dec.)
MDL Number: MFCD00012828
CAS Number: 19524-06-2
EINECS: 243-128-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR