3-Methylthiophene ≥98%

Supplier: BeanTown Chemical

218600-100G 218600-25G 218600-500G
BT218600-100GEA 115 USD
BT218600-100G BT218600-25G BT218600-500G
3-Methylthiophene ≥98%
3-Methylthiophene

CAS: 616-44-4; EC No: 210-482-6; MDL No: MFCD00005470; RTECS: XM9800000 UN No: UN1993; Haz Class: 3; Packing Group: II Liquid; Molecular Formula: C5H6S; MW: 98.17 Melting Point: -69°; Boiling Point: 114°/738 mmHg; Flash point: 12°C (54°F) Density (g/mL): 1.016; Refractive Index: 1.519


  • RTECS: XM9800000
  • Refractive index: 1.519
  • Liquid

Formula: C₅H₆S
Boiling Pt: 114 °C/738 mmHg
Melting Pt: –69 °C
Density: 1.016 g/mL
Flash Pt: 12 °C (54 °F)
MDL Number: MFCD00005470
CAS Number: 616-44-4
EINECS: 210-482-6
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR