3-Methylthiophene ≥98%
Supplier: BeanTown Chemical
218600-100G
218600-25G
218600-500G
BT218600-100GEA
115
USD
BT218600-100G
BT218600-25G
BT218600-500G
3-Methylthiophene ≥98%
3-Methylthiophene
CAS: 616-44-4; EC No: 210-482-6; MDL No: MFCD00005470; RTECS: XM9800000
UN No: UN1993; Haz Class: 3; Packing Group: II
Liquid; Molecular Formula: C5H6S; MW: 98.17
Melting Point: -69°; Boiling Point: 114°/738 mmHg; Flash point: 12°C (54°F)
Density (g/mL): 1.016; Refractive Index: 1.519
- RTECS: XM9800000
- Refractive index: 1.519
- Liquid
Formula:
C₅H₆S Boiling Pt: 114 °C/738 mmHg Melting Pt: –69 °C Density: 1.016 g/mL Flash Pt: 12 °C (54 °F) |
MDL Number:
MFCD00005470 CAS Number: 616-44-4 EINECS: 210-482-6 UN: 1993 ADR: 3,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...