Lead(II) nitrate ≥99%
Supplier: BeanTown Chemical
CAS: 10099-74-8; EC No: 233-245-9; MDL No: MFCD00011153; RTECS: OG2100000
UN No: UN1469; Haz Class: 5.1 (6.1); Packing Group: II
Powder; Linear Formula: Pb(NO3)2; MW: 331.20
Melting Point: 470° (decomposes)
Density (g/mL): 4.53
Hygroscopic
- RTECS: OG2100000
- Hygroscopic
- Powder
Formula:
Pb(NO₃)₂ Melting Pt: 470 °C (dec.) Density: 4.53 g/mL |
MDL Number:
MFCD00011153 CAS Number: 10099-74-8 EINECS: 233-245-9 UN: 1469 ADR: 5.1,II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...