2-Bromo-5-fluoropyridine ≥98%
Supplier: BeanTown Chemical
218470-1G
218470-5G
BT218470-1GEA
38
USD
BT218470-1G
BT218470-5G
2-Bromo-5-fluoropyridine ≥98%
2-Bromo-5-fluoropyridine
CAS: 41404-58-4; MDL No: MFCD00234011
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Solid; Molecular Formula: C5H3BrFN; MW: 175.99
Melting Point: 30-31°; Boiling Point: 80-83°/44 mmHg; Flash point: 73°C (163°F)
- Solid
Formula:
C₅H₃BrFN Boiling Pt: 80…83 °C/44 mmHg Melting Pt: 30…31 °C Flash Pt: 73 °C (163 °F) |
MDL Number:
MFCD00234011 CAS Number: 41404-58-4 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...