3-[(Trifluoromethyl)thio]aniline ≥96%

Supplier: BeanTown Chemical

218440-1G 218440-5G
BT218440-1GEA 89 USD
BT218440-1G BT218440-5G
3-[(Trifluoromethyl)thio]aniline ≥96%
3-[(Trifluoromethyl)thio]aniline

CAS: 369-68-6; EC No: 206-722-4; MDL No: MFCD00190130 UN No: UN2810; Haz Class: 6.1; Packing Group: III Liquid; Molecular Formula: C7H6F3NS ; MW: 193.19 Density (g/mL): 1.348


  • Liquid

Formula: C₇H₆F₃NS
Density: 1.348 g/mL
MDL Number: MFCD00190130
CAS Number: 369-68-6
EINECS: 206-722-4
ADR: 6.1,III

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR