1,2-Epoxy-4-vinylcyclohexane (mixture of isomers) ≥98%
Supplier: BeanTown Chemical
Synonyms:
3-Vinyl-7-oxabicyclo[4.1.0]heptane, 4-Vinyl-1,2-epoxycyclohexane
218425-250ML
BT218425-250MLEA
89.64
USD
BT218425-250ML
1,2-Epoxy-4-vinylcyclohexane (mixture of isomers) ≥98%
1,2-Epoxy-4-vinylcyclohexane (mixture of isomers)
CAS: 106-86-5; EC No: 203-436-1; MDL No: MFCD00022356; RTECS: RN8770000
UN No: UN3271; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C8H12O; MW: 124.18
Melting Point: -100°; Boiling Point: 169°; Flash point: 46°C (115°F)
Density (g/mL): 0.952; Refractive Index: 1.469
- RTECS: RN8770000
- Refractive index: 1.469
- Liquid
Formula:
C₈H₁₂O Boiling Pt: 169 °C Melting Pt: –100 °C Density: 0.952 g/mL Flash Pt: 46 °C (115 °F) |
MDL Number:
MFCD00022356 CAS Number: 106-86-5 EINECS: 203-436-1 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...