Magnesium acrylate
Supplier: BeanTown Chemical
218410-100G
218410-25G
218410-5G
BT218410-100GEA
271
USD
BT218410-100G
BT218410-25G
BT218410-5G
Magnesium acrylate
Magnesium acrylate
CAS: 5698-98-6; EC No: 227-177-9; MDL No: MFCD00068285
Powder; Linear Formula: (CH2=CHCOO)2Mg; Molecular Formula: C6H6MgO4 ; MW: 166.42
Melting Point:
- Air Sensitive, Moisture Sensitive
- Powder
Formula:
C₆H₆MgO₄ Melting Pt: >220 °C (dec.) |
MDL Number:
MFCD00068285 CAS Number: 5698-98-6 EINECS: 227-177-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...