Potassium iodate ≥98%
Supplier: BeanTown Chemical
218390-100G
218390-2.5KG
218390-500G
BT218390-100GEA
45
USD
BT218390-100G
BT218390-2.5KG
BT218390-500G
Potassium iodate ≥98%
Potassium iodate
CAS: 7758-05-6; EC No: 231-831-9; MDL No: MFCD00011406; RTECS: NN1350000
UN No: UN1479; Haz Class: 5.1; Packing Group: II
Crystalline/Powder; Linear Formula: KIO3; MW: 214.00
Melting Point: 560°
Density (g/mL): 3.93
- RTECS: NN1350000
- Crystalline/Powder
Formula:
KlO₃ MW: 214 g/mol Melting Pt: 560 Density: 3.93 g/mL |
MDL Number:
MFCD00011406 CAS Number: 7758-05-6 EINECS: 231-831-9 UN: 1479 ADR: 5.1 II Merck Index: 13,07726 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...