Tin(II) pyrophosphate
Supplier: BeanTown Chemical
Synonyms:
Stannous pyrophosphate
218385-250G
218385-50G
BT218385-250GEA
98
USD
BT218385-250G
BT218385-50G
Tin(II) pyrophosphate
Tin(II) pyrophosphate
CAS: 15578-26-4; EC No: 239-635-5; MDL No: MFCD00049544
Powder; Linear Formula: Sn2P2O7; MW: 411.32
Melting Point: 400° (decomposes)
Density (g/mL): 4.009
- Powder
Formula:
O₇P₂Sn₂ Melting Pt: 400 °C (dec.) Density: 4.009 g/mL |
MDL Number:
MFCD00049544 CAS Number: 15578-26-4 EINECS: 239-635-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...