Tricyclohexylphosphine ≥96%
Supplier: BeanTown Chemical
218310-1G
218310-25G
218310-5G
BT218310-1GEA
34.61
USD
BT218310-1G
BT218310-25G
BT218310-5G
Tricyclohexylphosphine ≥96%
Tricyclohexylphosphine
CAS: 2622-14-2; EC No: 220-069-2; MDL No: MFCD00003853
Crystalline/Powder; Linear Formula: (C6H11)3P ; MW: 280.42
Melting Point: 81-83°
Air Sensitive
- Air Sensitive
- Crystalline/Powder
Formula:
C₁₈H₃₃P Melting Pt: 81…83 °C |
MDL Number:
MFCD00003853 CAS Number: 2622-14-2 EINECS: 220-069-2 |
Specification Test Results
Assay | 96% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...