N-Acetylglycine(Ac-Gly-OH) ≥99%
Supplier: BeanTown Chemical
Synonyms:
Ac-Gly-OH, N-Acetylglycine, Acetamidoacetic acid, Aceturic acid
218295-100G
218295-500G
BT218295-100GEA
28
USD
BT218295-100G
BT218295-500G
N-Acetylglycine(Ac-Gly-OH) ≥99%
N-Acetylglycine (Ac-Gly-OH)
CAS: 543-24-8; EC No: 208-839-6; MDL No: MFCD00004275
Powder; Linear Formula: CH3CONHCH2COOH; Molecular Formula: C4H7NO3; MW: 117.10
Melting Point: 207-209°
- Powder
Formula:
C₄H₇NO₃ Melting Pt: 207…209 °C |
MDL Number:
MFCD00004275 CAS Number: 543-24-8 EINECS: 208-839-6 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...