Sodium bromide ≥99.99%
Supplier: BeanTown Chemical
218120-25G
218120-5G
BT218120-25GEA
192.99
USD
BT218120-25G
BT218120-5G
Sodium bromide ≥99.99%
Sodium bromide
CAS: 7647-15-6; EC No: 231-599-9; MDL No: MFCD00003475; RTECS: VZ3150000
Crystalline; Linear Formula: NaBr; MW: 102.89
Melting Point: 755°; Boiling Point: 1393°
Density (g/mL): 3.203; Refractive Index: 1.641
Hygroscopic
- RTECS: VZ3150000
- Refractive index: 1.641
- Hygroscopic
- Crystalline
Formula:
NaBr Boiling Pt: 1393 °C Melting Pt: 755 °C Density: 3.203 g/mL |
MDL Number:
MFCD00003475 CAS Number: 7647-15-6 EINECS: 231-599-9 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...