2-Chloro-2-methylhexane ≥97%

Supplier: BeanTown Chemical

218035-25G 218035-5G
BT218035-25GEA 192.91 USD
BT218035-25G BT218035-5G
2-Chloro-2-methylhexane ≥97%
2-Chloro-2-methylhexane

CAS: 4398-65-6; MDL No: MFCD00060779 UN No: UN1993; Haz Class: 3; Packing Group: III Liquid; Linear Formula: CH3(CH2)3CCl(CH3)2; Molecular Formula: C7H15Cl; MW: 134.65


  • Liquid

Formula: C₇H₁₅Cl
MDL Number: MFCD00060779
CAS Number: 4398-65-6
ADR: 3,III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR