4-Methoxypyridine ≥98%

Supplier: BeanTown Chemical

217955-25G 217955-5G
BT217955-25GEA 231 USD
BT217955-25G BT217955-5G
4-Methoxypyridine ≥98%
4-Methoxypyridine

CAS: 620-08-6; EC No: 210-624-7; MDL No: MFCD00674049 Liquid; Molecular Formula: C6H7NO ; MW: 109.13 Boiling Point: 108-111°/65 mmHg; Flash point: 74°C (165°F) Density (g/mL): 1.075 ; Refractive Index: 1.516


  • Refractive index: 1.516
  • Liquid

Formula: C₆H₇NO
Boiling Pt: 108…111 °C/65 mmHg
Density: 1.075 g/mL
Flash Pt: 74 °C (165 °F)
MDL Number: MFCD00674049
CAS Number: 620-08-6
EINECS: 210-624-7

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR