trans-β-Nitrostyrene ≥98%
Supplier: BeanTown Chemical
217940-100G
217940-25G
217940-5G
BT217940-100GEA
290
USD
BT217940-100G
BT217940-25G
BT217940-5G
trans-β-Nitrostyrene ≥98%
trans-β-Nitrostyrene
CAS: 5153-67-3; EC No: 203-066-0; MDL No: MFCD00007402; RTECS: WL5460000
Crystalline; Linear Formula: C6H5CH=CHNO2; Molecular Formula: C8H7NO2; MW: 149.15
Melting Point: 55-58°; Boiling Point: 250-260°
- RTECS: WL5460000
- Crystalline
Formula:
C₈H₇NO₂ Boiling Pt: 250…260 °C Melting Pt: 55…58 °C |
MDL Number:
MFCD00007402 CAS Number: 5153-67-3 EINECS: 203-066-0 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...