Copper(II) oxide ≥99.0% ACS
Supplier: BeanTown Chemical
217935-1KG
217935-250G
BT217935-1KGEA
188
USD
BT217935-1KG
BT217935-250G
Copper(II) oxide ≥99.0% ACS
Copper (II) oxide
CAS: 1317-38-0; EC No: 215-269-1; MDL No: MFCD00010979; RTECS: GL7900000
UN No: UN3077; Haz Class: 9; Packing Group: III
Powder; Molecular Formula: CuO; MW: 79.55
Melting Point: 1330°
Density (g/mL): 6.4
- RTECS: GL7900000
- Powder
Formula:
CuO Melting Pt: 1330 °C Density: 6.4 g/mL |
MDL Number:
MFCD00010979 CAS Number: 1317-38-0 EINECS: 215-269-1 UN: 3077 ADR: 9,III |
Specification Test Results
Assay | 99.0% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...