Nickel(II) stearate
Supplier: BeanTown Chemical
Synonyms:
Nickel stearate
217870-100G
217870-2KG
217870-500G
BT217870-100GEA
40
USD
BT217870-100G
BT217870-2KG
BT217870-500G
Nickel(II) stearate
Nickel (II) stearate
CAS: 2223-95-2; EC No: 218-744-1; MDL No: MFCD00054344
UN No: UN3077; Haz Class: 9; Packing Group: III
Powder; Linear Formula: [CH3(CH2)16COO]2Ni; Molecular Formula: C36H70NiO4; MW: 625.63
Melting Point: 82-86°
Density (g/mL): 1.13
- Powder
Formula:
C₃₆H₇₀NiO₄ Melting Pt: 82…86 °C Density: 1.13 g/mL |
MDL Number:
MFCD00054344 CAS Number: 2223-95-2 EINECS: 218-744-1 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...