Sodium 2,4-dinitrobenzenesulfonate ≥97%

Supplier: BeanTown Chemical

Synonyms: 2,4-Dinitrobenzenesulfonic acid sodium salt, 2,4-Dinitrobenzenesulphonic acid sodium salt

217820-25G 217820-5G
BT217820-25GEA 72 USD
BT217820-25G BT217820-5G
Sodium 2,4-dinitrobenzenesulfonate ≥97%
Sodium 2,4-dinitrobenzenesulphonate

CAS: 885-62-1; EC No: 212-943-7; MDL No: MFCD00007471 Powder; Molecular Formula: C6H3N2NaO7S; MW: 270.15


  • Powder

Formula: C₆H₃N₂NaO₇S
MDL Number: MFCD00007471
CAS Number: 885-62-1
EINECS: 212-943-7

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR