Strontium chloride, anhydrous ≥95%

Supplier: BeanTown Chemical

217800-2KG 217800-500G
BT217800-2KGEA 160 USD
BT217800-2KG BT217800-500G
Strontium chloride, anhydrous ≥95%
Strontium chloride

CAS: 10476-85-4; EC No: 233-971-6; MDL No: MFCD00011249 Powder; Linear Formula: SrCl2; MW: 158.53 Melting Point: 873°; Boiling Point: 1250° Density (g/mL): 3.052; Refractive Index: 1.65 Hygroscopic


  • Refractive index: 1.65
  • Hygroscopic
  • Powder

Formula: Cl₂Sr
MW: 158.53
Boiling Pt: 1250
Melting Pt: 868
Density: 3.052 g/mL
MDL Number: MFCD00011249
CAS Number: 10476-85-4
EINECS: 233-971-6
Merck Index: 12,09000

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR