Hexamethyldisiloxane ≥99.5%
Supplier: BeanTown Chemical
Synonyms:
Silicone fluid, HMDSO
217705-100G
217705-25G
BT217705-100GEA
69.2
USD
BT217705-100G
BT217705-25G
Hexamethyldisiloxane ≥99.5%
Hexamethyldisiloxane
CAS: 107-46-0; EC No: 203-492-7; MDL No: MFCD00008265; RTECS: JM9237000
UN No: UN1993; Haz Class: 3; Packing Group: II
Liquid; Linear Formula: (CH3)3SiOSi(CH3)3; Molecular Formula: C6H18OSi2; MW: 162.38
Melting Point: -59°; Boiling Point: 101°; Flash point: -2°C (28°F)
Density (g/mL): 0.764; Refractive Index: 1.377
- RTECS: JM9237000
- Refractive index: 1.377
- Liquid
Formula:
C₆H₁₈OSi₂ Boiling Pt: 101 °C Melting Pt: –59 °C Density: 0.764 g/mL Flash Pt: –2 °C (28 °F) |
MDL Number:
MFCD00008265 CAS Number: 107-46-0 EINECS: 203-492-7 UN: 1993 ADR: 3,II |
|
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...