Iron(III) nitrate nonahydrate 98.0-101.0% ACS

Supplier: BeanTown Chemical

Synonyms: Ferric nitrate nonahydrate, Iron trinitrate nonahydrate, Iron(III) nitrate hydrate, Ferric Nitrate, 9-Hydrate

217660-2KG 217660-500G
BT217660-2KGEA 185.62 USD
BT217660-2KG BT217660-500G
Iron(III) nitrate nonahydrate 98.0-101.0% ACS
Iron (III) nitrate nonahydrate

CAS: 7782-61-8; EC No: 233-899-5; MDL No: MFCD00149708; RTECS: NO7175000 UN No: UN1466; Haz Class: 5.1; Packing Group: III Crystalline; Linear Formula: Fe(NO3)3·9H2O; MW: 403.99 Melting Point: 47°; Boiling Point: 125° (decomposes) Moisture Sensitive


  • RTECS: NO7175000
  • Moisture Sensitive
  • Crystalline

Formula: Fe(NO₃)₃·9H₂O
MW: 404 g/mol
Boiling Pt: 125 (dec.)
Melting Pt: 47
MDL Number: MFCD00149708
CAS Number: 7782-61-8
EINECS: 233-899-5
UN: 1466
ADR: 5.1 III
Merck Index: 13,04057

Order Now

Specification Test Results

Assay 98.0-101.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR