Praseodymium ≥99% (trace metals basis excluding Ta), rods Ø 6.35 mm

Supplier: BeanTown Chemical

217640-10MM 217640-25MM 217640-50MM
BT217640-10MMEA 130 USD
BT217640-10MM BT217640-25MM BT217640-50MM
Praseodymium ≥99% (trace metals basis excluding Ta), rods Ø 6.35 mm
Praseodymium

CAS: 7440-10-0; EC No: 231-120-3; MDL No: MFCD00011174 Solid; Molecular Formula: Pr; MW: 140.91 Melting Point: 931°; Boiling Point: 3520° Density (g/mL): 6.71


  • Solid

Formula: Pr
MW: 140.91
Boiling Pt: 3520 °C
Melting Pt: 931 °C
Density: 6.71 g/mL
MDL Number: MFCD00011174
CAS Number: 7440-10-0
EINECS: 231-120-3

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR