1-Pentanol ≥99.0% ACS
Supplier: BeanTown Chemical
Synonyms:
1-Pentyl alcohol, 1-Hydroxy pentane, n-Amyl alcohol, Amyl alcohol, Butyl carbinol
217605-1L
217605-250ML
217605-4L
BT217605-1LEA
61.78
USD
BT217605-1L
BT217605-250ML
BT217605-4L
1-Pentanol ≥99.0% ACS
1-Pentanol
CAS: 71-41-0; EC No: 200-752-1; MDL No: MFCD00002977; RTECS: SB9800000
UN No: UN1105; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: CH3(CH2)4OH; Molecular Formula: C5H12O ; MW: 88.15
Melting Point: -78°; Boiling Point: 136-138°; Flash point: 49°C (120°F)
Density (g/mL): 0.811; Refractive Index: 1.409
- RTECS: SB9800000
- Refractive index: 1.409
- Liquid
Formula:
H₃C(CH₂)₃CH₂OH Boiling Pt: 136…138 °C Melting Pt: –78 °C Density: 0.811 g/mL Flash Pt: 49 °C (120 °F) |
MDL Number:
MFCD00002977 CAS Number: 71-41-0 EINECS: 200-752-1 UN: 1105 ADR: 3,III |
|
Specification Test Results
Assay | 99.0% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...