Methyl purple 0,1% (w/v) in water

Supplier: BeanTown Chemical

Synonyms: Fleisher's Indicator Solution

217580-1L 217580-500ML
BT217580-1LEA 47 USD
BT217580-1L BT217580-500ML
Methyl purple 0,1% (w/v) in water
Methyl purple

CAS: 1340-02-9; MDL No: MFCD00070843 Liquid; Molecular Formula: C67H62N8Na4O14S3; MW: 1391.41 Melting Point: 0°; Boiling Point: 100° Density (g/mL): 1.001


  • Liquid

Formula: C₆₇H₆₂N₈Na₄O₁₄S₃
MW: 1391.41 g/mol
Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 1.001 g/mL
MDL Number: MFCD00070843
CAS Number: 1340-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR