2-chloro-5-iodopyrimidine ≥95%

Supplier: BeanTown Chemical

Synonyms: 2-Chloro-5-iodopyrimidine

217475-1G 217475-250MG 217475-5G
BT217475-1GEA 246 USD
BT217475-1G BT217475-250MG BT217475-5G
2-chloro-5-iodopyrimidine ≥95%
2-chloro-5-iodopyrimidine

CAS: 32779-38-7 Solid; Molecular Formula: C4H2ClIN2; MW: 240.43 Light Sensitive


  • Light Sensitive
  • Solid

Formula: C₄H₂ClIN₂
MDL Number: MFCD06200210
CAS Number: 32779-38-7

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR