Tetraethylammonium tetrafluoroborate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Tetrylammonium tetrafluoroborate
217455-100G
217455-50G
217455-5G
BT217455-100GEA
247.54
USD
BT217455-100G
BT217455-50G
BT217455-5G
Tetraethylammonium tetrafluoroborate ≥99%
Tetraethylammonium tetrafluoroborate
CAS: 429-06-1; EC No: 207-055-1; MDL No: MFCD00011827
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Solid; Linear Formula: (CH3CH2)4N(BF4); Molecular Formula: C8H20BF4N; MW: 217.06
Melting Point: 370-380° (decomposes)
Hygroscopic
- Hygroscopic
- Solid
Formula:
C₈H₂₀BF₄N Melting Pt: 370…380 °C (dec.) |
MDL Number:
MFCD00011827 CAS Number: 429-06-1 EINECS: 207-055-1 UN: 2811 ADR: 6.1,III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...