Tantalum ≥99.9% (trace metals basis), powder APS ∼2 µm
Supplier: BeanTown Chemical
217450-25G
217450-50G
217450-5G
BT217450-25GEA
204
USD
BT217450-25G
BT217450-50G
BT217450-5G
Tantalum ≥99.9% (trace metals basis), powder APS ∼2 µm
Tantalum
CAS: 7440-25-7; EC No: 231-135-5; MDL No: MFCD00011252; RTECS: WW5505000
UN No: UN3089; Haz Class: 4.1; Packing Group: II
Powder; Molecular Formula: Ta; MW: 180.95
Melting Point: 2996°; Boiling Point: 5425°
Density (g/mL): 16.69
- RTECS: WW5505000
- Powder
Formula:
Ta MW: 180.95 g/mol Boiling Pt: 5427 Melting Pt: 2996 Density: 16.69 g/mL |
MDL Number:
MFCD00011252 CAS Number: 7440-25-7 EINECS: 231-135-5 UN: 3089 ADR: 4.1 II Merck Index: 12,09223 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...