D-Methionine ≥99%

Supplier: BeanTown Chemical

Synonyms: D(+)-Methionine, (R)-2-Amino-4-(methylthio)butanoic acid, (R)-(+)-2-Amino-4-(methylthio)butyric acid, (R)-(+)-2-Amino-4-(methylthio)butanoic acid, (R)-(+)-Methionine, H-D-Met-OH

217445-100G 217445-25G 217445-5G
BT217445-100GEA 310 USD
BT217445-100G BT217445-25G BT217445-5G
D-Methionine ≥99%
D-Methionine

CAS: 348-67-4; EC No: 206-483-6; MDL No: MFCD00002622; RTECS: PD0455000 Crystalline/Powder; Molecular Formula: C5H11NO2S; MW: 149.21 Optical Rotation: [α]20/D -23.7±0.5°, c = 5% in 5M HCl


  • RTECS: PD0455000
  • Optical rotation: [α]20/D -23.7±0.5°, c = 5% in 5M HCl
  • Crystalline/Powder

Formula: C₅H₁₁NO₂S
MDL Number: MFCD00002622
CAS Number: 348-67-4
EINECS: 206-483-6

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR