Zirconyl nitrate hydrate ≥99.9%

Supplier: BeanTown Chemical

Synonyms: Zirconium(IV) oxynitrate hydrate, Zirconium(IV) dinitrate oxide hydrate, Zirconium dinitrate oxide hydrate

217385-100G 217385-25G 217385-500G
BT217385-100GEA 207.71 USD
BT217385-100G BT217385-25G BT217385-500G
Zirconyl nitrate hydrate ≥99.9%
Zirconyl nitrate hydrate

CAS: 14985-18-3; EC No: 237-529-3; MDL No: MFCD00149896 UN No: UN1477; Haz Class: 5.1; Packing Group: III Crystalline; Linear Formula: ZrO(NO3)2·xH2O; MW: 231.23 (anhydrous)


  • Crystalline

Formula: N₂O₇Zr·H₂O
MDL Number: MFCD00149896
CAS Number: 14985-18-3
EINECS: 237-529-3
UN: 1477
ADR: 5.1,III

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR