Antimony ≥99.999%, shot 6 mm

Supplier: BeanTown Chemical

217180-100G 217180-25G 217180-500G
BT217180-100GEA 245 USD
BT217180-100G BT217180-25G BT217180-500G
Antimony ≥99.999%, shot 6 mm
Antimony

CAS: 7440-36-0; EC No: 231-146-5; MDL No: MFCD00134030; RTECS: CC4025000 Solid; Molecular Formula: Sb; MW: 121.76 Melting Point: 630°; Boiling Point: 1635° Density (g/mL): 6.69


  • RTECS: CC4025000
  • Solid

Formula: Sb
MW: 121.76 g/mol
Boiling Pt: 1637
Melting Pt: 630
Density: 6.69 g/mL
MDL Number: MFCD00134030
CAS Number: 7440-36-0
EINECS: 231-146-5
UN: 2871
ADR: 6.1 III
Merck Index: 12,00733

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR