4-Hydroxy-3-methoxycinnamic acid (Ferulic acid) ≥99%
Supplier: BeanTown Chemical
217130-10G
BT217130-10GEA
34.2
USD
BT217130-10G
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid) ≥99%
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid)
CAS: 1135-24-6; EC No: 214-490-0; MDL No: MFCD00004400; RTECS: UD3365500
Powder; Molecular Formula: C10H10O4; MW: 194.18
Melting Point: 168-172°
- RTECS: UD3365500
- Powder
Formula:
C₁₀H₁₀O₄ Melting Pt: 168…172 °C |
MDL Number:
MFCD00004400 CAS Number: 1135-24-6 EINECS: 214-490-0 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...