Neocuproine hemihydrate ≥98%
Supplier: BeanTown Chemical
Synonyms:
2,9-Dimethyl-1,10-phenanthroline hemihydrate
217120-100G
217120-25G
217120-5G
BT217120-100GEA
847
USD
BT217120-100G
BT217120-25G
BT217120-5G
Neocuproine hemihydrate ≥98%
Neocuproine hemihydrate
CAS: 34302-69-7; EC No: 207-601-9; MDL No: MFCD00149306
Solid; Molecular Formula: C14H12N2·0.5H2O; MW: 217.27
Melting Point: 159-164°
- Solid
Formula:
C₂₈H₂₆N₄O MW: 434.53 g/mol Melting Pt: 159…164 °C |
MDL Number:
MFCD00149306 CAS Number: 34302-69-7 EINECS: 207-601-9 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...