Methyl cellulose, 1600 cPs (2% H₂O)

Supplier: BeanTown Chemical

217105-100G 217105-2.5KG 217105-500G
BT217105-100GEA 40.53 USD
BT217105-100G BT217105-2.5KG BT217105-500G
Methyl cellulose, 1600 cPs (2% H₂O)
Methyl cellulose

CAS: 9004-67-5; MDL No: MFCD00081763 Powder


  • Powder

MDL Number: MFCD00081763
CAS Number: 9004-67-5
Merck Index: 14,6040

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR