Methyl cellulose, 1600 cPs (2% H₂O)
Supplier: BeanTown Chemical
217105-100G
217105-2.5KG
217105-500G
BT217105-100GEA
40.53
USD
BT217105-100G
BT217105-2.5KG
BT217105-500G
Methyl cellulose, 1600 cPs (2% H₂O)
Methyl cellulose
CAS: 9004-67-5; MDL No: MFCD00081763
Powder
- Powder
MDL Number:
MFCD00081763 CAS Number: 9004-67-5 Merck Index: 14,6040 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...