(3-Bromo-4-methoxyphenyl)acetic acid

Supplier: BeanTown Chemical

Synonyms: 3-Bromo-4-methoxyphenylacetic acid, 2-(3-Bromo-4-methoxyphenyl)acetic acid

217085-1G 217085-5G
BT217085-1GEA 92 USD
BT217085-1G BT217085-5G
(3-Bromo-4-methoxyphenyl)acetic acid
(3-Bromo-4-methoxyphenyl)acetic acid

CAS: 774-81-2; MDL No: MFCD00017538 Solid; Molecular Formula: C9H9BrO3; MW: 245.07


  • Solid

Formula: C₉H₉BrO₃
MW: 245.07
MDL Number: MFCD00017538
CAS Number: 774-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR