3,5,5-Trimethylcyclohexane-1,2-dione ≥99%
Supplier: BeanTown Chemical
217065-25G
217065-5G
BT217065-25GEA
244
USD
BT217065-25G
BT217065-5G
3,5,5-Trimethylcyclohexane-1,2-dione ≥99%
3,5,5-Trimethylcyclohexane-1,2-dione
CAS: 57696-89-6; MDL No: MFCD02181023
Solid; Molecular Formula: C9H14O2 ; MW: 154.21
- Solid
Formula:
C₉H₁₄O₂ |
MDL Number:
MFCD02181023 CAS Number: 57696-89-6 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...