Platinum ≥99.99% (trace metals basis), powder -22 mesh

Supplier: BeanTown Chemical

Synonyms: Platinium

217030-1G 217030-250MG 217030-5G
BT217030-1GEA 292.35 USD
BT217030-1G BT217030-250MG BT217030-5G
Platinum ≥99.99% (trace metals basis), powder -22 mesh
Platinum

CAS: 7440-06-4; MDL No: MFCD00011179; RTECS: TP2160000 Powder; Molecular Formula: Pt; MW: 195.08 Melting Point: 1772°; Boiling Point: 3827° Density (g/mL): 21.45


  • RTECS: TP2160000
  • Powder

Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Density: 21.45 g/mL
MDL Number: MFCD00011179
CAS Number: 7440-06-4
UN: 3089
ADR: 4.1 II
Merck Index: 12,07689

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR