Benzyl acrylate ≥97% stabilised

Supplier: BeanTown Chemical

Synonyms: Acrylic Acid Benzyl Ester

216895-10G 216895-250G 216895-50G
BT216895-10GEA 27 USD
BT216895-10G BT216895-250G BT216895-50G
Benzyl acrylate ≥97% stabilised
Benzyl acrylate

CAS: 2495-35-4; EC No: 219-673-9; MDL No: MFCD00048147 UN No: UN3082; Haz Class: 9; Packing Group: III Liquid; Molecular Formula: C10H10O2; MW: 162.19 Boiling Point: 111°/8 mmHg Density (g/mL): 1.06; Refractive Index: 1.518


  • Refractive index: 1.518
  • Liquid

Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Boiling Pt: 111 °C/8 mmHg
Density: 1.06 g/mL
MDL Number: MFCD00048147
CAS Number: 2495-35-4
EINECS: 219-673-9
ADR: 9 III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR