Iodomethane ≥99% stabilised
Supplier: BeanTown Chemical
Synonyms:
Methyl iodide
216890-1KG
216890-250G
216890-50G
BT216890-1KGEA
307.88
USD
BT216890-1KG
BT216890-250G
BT216890-50G
Iodomethane ≥99% stabilised
Iodomethane
CAS: 74-88-4; EC No: 200-819-5; MDL No: MFCD00001073; RTECS: PA9450000
UN No: UN2644; Haz Class: 6.1; Packing Group: I
Liquid; Molecular Formula: CH3I; MW: 141.94
Melting Point: -64°; Boiling Point: 41-43°
Density (g/mL): 2.28; Refractive Index: 1.531
Light Sensitive
- RTECS: PA9450000
- Refractive index: 1.531
- Light Sensitive
- Liquid
Formula:
CH₃I Boiling Pt: 41…43 °C Melting Pt: –64 °C Density: 2.28 g/mL |
MDL Number:
MFCD00001073 CAS Number: 74-88-4 EINECS: 200-819-5 UN: 2644 ADR: 6.1,I |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...