(1S)-(+)-Camphor-10-sulfonyl chloride ≥97%

Supplier: BeanTown Chemical

Synonyms: (1S)-(+)-2-Oxobornane-10-sulfonyl chloride, (1S)-(+)-2-Oxobornane-10-sulphonyl chloride, D(+)-Camphor-10-sulfonyl chloride, ((1S,4R)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulphonyl chloride, ((1S,4R)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulfonyl chloride, D(+)-Camphor-10-sulphonyl chloride

216885-100G 216885-25G 216885-5G
BT216885-100GEA 342 USD
BT216885-100G BT216885-25G BT216885-5G
(1S)-(+)-Camphor-10-sulfonyl chloride ≥97%
(1S)-(+)-Camphor-10-sulphonyl chloride

CAS: 21286-54-4; MDL No: MFCD00064156 UN No: UN3261; Haz Class: 8; Packing Group: II Crystalline; Molecular Formula: C10H15ClO3S ; MW: 250.74 Melting Point: 65-67° Optical Rotation: [α]22/D +33°, c = 1 in chloroform Moisture Sensitive


  • Optical rotation: [α]22/D +33°, c = 1 in chloroform
  • Moisture Sensitive
  • Crystalline

Formula: C₁₀H₁₅ClO₃S
MW: 250.75
Melting Pt: 70
MDL Number: MFCD00064156
CAS Number: 21286-54-4
UN: 3261
ADR: 8 II

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR