Cresol red sodium salt

Supplier: BeanTown Chemical

Synonyms: Cresol Red sodium, Sodium 4-(3-(4-hydroxy-3-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-2-methylphenolate

216835-100G 216835-25G 216835-5G
BT216835-100GEA 176 USD
BT216835-100G BT216835-25G BT216835-5G
Cresol red sodium salt
Cresol red sodium salt

CAS: 62625-29-0; EC No: 263-654-8; MDL No: MFCD00001618 Powder; Molecular Formula: C21H17NaO5S; MW: 404.42 Melting Point: 250° (decomposes)


  • Powder

Formula: C₂₁H₁₇NaO₅S
Melting Pt: 250 °C (dec.)
MDL Number: MFCD00001618
CAS Number: 62625-29-0
EINECS: 263-654-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR