Methyl oxalyl chloride ≥97%

Supplier: BeanTown Chemical

Synonyms: Methyl 2-chloro-2-oxoacetate, Methyl chloroglyoxylate

216780-100G 216780-25G 216780-5G
BT216780-100GEA 282 USD
BT216780-100G BT216780-25G BT216780-5G
Methyl oxalyl chloride ≥97%
Methyl oxalyl chloride

CAS: 5781-53-3; EC No: 227-307-4; MDL No: MFCD00000705 UN No: UN2920; Haz Class: 8 (3); Packing Group: II Liquid; Linear Formula: ClCOCOOCH3; Molecular Formula: C3H3ClO3; MW: 122.51 Boiling Point: 118-120°; Flash point: 46°C (114°F) Density (g/mL): 1.332; Refractive Index: 1.419 Moisture Sensitive


  • Refractive index: 1.419
  • Moisture Sensitive
  • Liquid

Formula: C₃H₃ClO₃
MW: 122.51 g/mol
Boiling Pt: 118…120 °C
Density: 1.332 g/mL
Flash Pt: 46 °C (114 °F)
MDL Number: MFCD00000705
CAS Number: 5781-53-3
EINECS: 227-307-4
UN: 2920
ADR: 8 II

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR