Rhodamine B
Supplier: BeanTown Chemical
Synonyms:
9-(2-Carboxyphenyl)-3,6-bis(diethylamino)xanthylium chloride, [9-(2-Carboxyphenyl)-6-(diethylamino)xanthen-3-ylidene]-diethylazanium chloride
216735-1KG
216735-250G
216735-50G
BT216735-1KGEA
225
USD
BT216735-1KG
BT216735-250G
BT216735-50G
Rhodamine B
Rhodamine B
CAS: 81-88-9; EC No: 201-383-9; MDL No: MFCD00011931; RTECS: BP3675000
Powder; Molecular Formula: C28H31ClN2O3; MW: 479.02
Melting Point: 210-211° (decomposes)
Density (g/mL): 0.790
- RTECS: BP3675000
- Powder
Formula:
C₂₈H₃₁ClN₂O₃ Melting Pt: 210…211 °C (dec.) Density: 0.790 g/mL |
MDL Number:
MFCD00011931 CAS Number: 81-88-9 EINECS: 201-383-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...