N,N-Dimethylformamide-[D₇] ≥99.5% (Isotopic)
Supplier: BeanTown Chemical
Synonyms:
Heptadeutero-N,N-dimethylformamide, N,N-Di[2H3]methyl[2H]formamide, N,N-Dimethylformamide-d7, DMF-D7, DMF-d7, N,N-Dimethylformamide-[²H₇], Formdimethylamide-D7, Formic acid dimethylamide-D7
216675-1G
216675-5G
216675-5X1G
BT216675-1GEA
123.33
USD
BT216675-1G
BT216675-5G
BT216675-5X1G
N,N-Dimethylformamide-[D₇] ≥99.5% (Isotopic)
N,N-Dimethylformamide-[D7]
CAS: 4472-41-7; EC No: 224-745-8; MDL No: MFCD00003286
UN No: UN2265; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: DCON(CD3)2; Molecular Formula: C3D7NO; MW: 80.14
Boiling Point: 153°; Flash point: 58°C (136°F)
Density (g/mL): 1.03; Refractive Index: 1.428
Hygroscopic
- Refractive index: 1.428
- Hygroscopic
- Liquid
Formula:
DCOON(CD₃)₂ Boiling Pt: 153 °C Density: 1.03 g/mL Flash Pt: 58 °C (136 °F) |
MDL Number:
MFCD00003286 CAS Number: 4472-41-7 EINECS: 224-745-8 UN: 2265 ADR: 3,III |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...