Chromium(VI) oxide ≥98.0% ACS
Supplier: BeanTown Chemical
Synonyms:
Chromic anhydride, Chromium trioxide
216635-100G
216635-500G
BT216635-100GEA
70.53
USD
BT216635-100G
BT216635-500G
Chromium(VI) oxide ≥98.0% ACS
Chromium (VI) oxide
CAS: 1333-82-0; EC No: 215-607-8; MDL No: MFCD00010952; RTECS: GB6650000
UN No: UN1463; Haz Class: 5.1 (6.1,8); Packing Group: II
Crystalline; Molecular Formula: CrO3; MW: 99.99
Melting Point: 196° (decomposes)
Density (g/mL): 2.7
Hygroscopic
- RTECS: GB6650000
- Hygroscopic
- Crystalline
Formula:
CrO₃ Melting Pt: 196 °C (dec.) Density: 2.7 g/mL |
MDL Number:
MFCD00010952 CAS Number: 1333-82-0 EINECS: 215-607-8 UN: 1463 ADR: 5.1,II |
Specification Test Results
Assay | 98.0% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...